COMH4211D21B Combi-Blocks, Inc.

2-Chloro-5-(2-methoxyphenyl)pyrimidine

  • MDL number MFCD19318522
  • Cas Number : 1267009-84-6
  • Molecular Weight : 220.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9658

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9658
Availability details for
Quantity
  • Availability
  • Add to Cart