COMH4211D21D Combi-Blocks, Inc.

5-Bromo-2-chloro-nicotinic acid tert-butyl ester

  • MDL number MFCD18897599
  • Cas Number : 1335055-73-6
  • Molecular Weight : 292.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9663

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9663
Availability details for
Quantity
  • Availability
  • Add to Cart