COMH4211D21F Combi-Blocks, Inc.

1-Phenyl-4-(prop-2-en-1-yl)piperazine

  • MDL number MFCD12827849
  • Cas Number : 21057-43-2
  • Molecular Weight : 202.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9667

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9667
Availability details for
Quantity
  • Availability
  • Add to Cart