COMH4211D220 Combi-Blocks, Inc.

1-Bromo-4-(cyclopropylmethoxy)-2-fluorobenzene

  • MDL number MFCD20882276
  • Cas Number : 1353776-86-9
  • Molecular Weight : 245.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9670

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9670
Availability details for
Quantity
  • Availability
  • Add to Cart