COMH4211D222 Combi-Blocks, Inc.

3-(2-(2-[2-(Tetrahydro-pyran-2-yloxy)-ethoxy]-ethoxy)-ethoxy)-propionic acid

  • MDL number MFCD31381618
  • Cas Number : 2203140-34-3
  • Molecular Weight : 306.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9678

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9678
Availability details for
Quantity
  • Availability
  • Add to Cart