COMH4211D227 Combi-Blocks, Inc.

1-(4-(Methylsulfonyl)benzyl)piperazine

  • MDL number MFCD11590753
  • Cas Number : 1082399-22-1
  • Molecular Weight : 254.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9687

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9687
Availability details for
Quantity
  • Availability
  • Add to Cart