COMH4211D229 Combi-Blocks, Inc.

Ethyl 2-(2-chloro-6-methylphenyl)-2,2-difluoroacetate

  • MDL number MFCD31579423
  • Cas Number : 1912447-92-7
  • Molecular Weight : 248.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9690

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9690
Availability details for
Quantity
  • Availability
  • Add to Cart