COMH4211D22A Combi-Blocks, Inc.

3-Methyl-4-(pyrrolidine-1-carbonyl)aniline

  • MDL number MFCD16117672
  • Cas Number : 325125-06-2
  • Molecular Weight : 204.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9692

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9692
Availability details for
Quantity
  • Availability
  • Add to Cart