COMH4211D230 Combi-Blocks, Inc.

S-[(Trans-4-aminocyclohexyl)methyl]ethanethioate trifluoroacetate

  • MDL number MFCD31579511
  • Cas Number : 1607590-28-2
  • Molecular Weight : 301.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9705

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9705
Availability details for
Quantity
  • Availability
  • Add to Cart