COMH4211D236 Combi-Blocks, Inc.

4-Amino-3-(cyclopropylmethoxy)benzamide

  • MDL number MFCD18338364
  • Cas Number : 1339747-63-5
  • Molecular Weight : 206.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9729

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9729
Availability details for
Quantity
  • Availability
  • Add to Cart