COMH4211D238 Combi-Blocks, Inc.

5-Fluoro-3-n-hexyloxybenzoic acid

  • MDL number MFCD18426790
  • Cas Number : 1443333-60-5
  • Molecular Weight : 240.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9732

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9732
Availability details for
Quantity
  • Availability
  • Add to Cart