COMH4211D239 Combi-Blocks, Inc.

1-(2-Bromoethoxy)-4-fluoro-2-(trifluoromethyl)benzene

  • MDL number MFCD27946855
  • Cas Number : 194876-78-3
  • Molecular Weight : 287.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9734

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9734
Availability details for
Quantity
  • Availability
  • Add to Cart