COMH4211D23B Combi-Blocks, Inc.

3-Amino-n,n-diethyl-5-fluorobenzamide

  • MDL number MFCD24125079
  • Cas Number : 1518386-29-2
  • Molecular Weight : 210.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9740

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9740
Availability details for
Quantity
  • Availability
  • Add to Cart