COMH4211D23C Combi-Blocks, Inc.

4-Hydroxy-2-methyl-n-(propan-2-yl)benzamide

  • MDL number MFCD31613839
  • Cas Number : 1692193-58-0
  • Molecular Weight : 193.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9747

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9747
Availability details for
Quantity
  • Availability
  • Add to Cart