COMH4211D23D Combi-Blocks, Inc.

2-((3-(Trifluoromethyl)pyridin-2-yl)oxy)ethan-1-ol

  • MDL number MFCD29046287
  • Cas Number : 170502-07-5
  • Molecular Weight : 207.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9749

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9749
Availability details for
Quantity
  • Availability
  • Add to Cart