COMH4211D23F Combi-Blocks, Inc.

4-[2-(1,3-Dioxo-1,3-dihydro-2h-isoindol-2-yl)ethyl]sulfanylbenzonitrile

  • MDL number MFCD31614046
  • Cas Number : 2270907-41-8
  • Molecular Weight : 308.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9753

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9753
Availability details for
Quantity
  • Availability
  • Add to Cart