COMH4211D24B Combi-Blocks, Inc.

5-Bromo-2-methyl-n-(propan-2-yl)benzamide

  • MDL number MFCD21193015
  • Cas Number : 1487693-85-5
  • Molecular Weight : 256.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9780

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9780
Availability details for
Quantity
  • Availability
  • Add to Cart