COMH4211D253 Combi-Blocks, Inc.

1-[3-Chloro-4-(2-methylpropoxy)phenyl]ethan-1-amine

  • MDL number MFCD16314599
  • Cas Number : 742002-96-6
  • Molecular Weight : 227.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9791

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9791
Availability details for
Quantity
  • Availability
  • Add to Cart