COMH4211D25A Combi-Blocks, Inc.

N-([2-(Trifluoromethyl)phenyl]methyl)cyclopropanamine

  • MDL number MFCD08060649
  • Cas Number : 920462-64-2
  • Molecular Weight : 215.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9799

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9799
Availability details for
Quantity
  • Availability
  • Add to Cart