COMH4211D25C Combi-Blocks, Inc.

1-(4-Methoxy-benzyl)-3,5-dimethyl-4-(4,4,5,5-tetramethyl-[1,3,2]dioxaborolan-2-yl)-1h-pyrazole

  • MDL number MFCD22572227
  • Cas Number : 1232027-73-4
  • Molecular Weight : 342.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9801

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9801
Availability details for
Quantity
  • Availability
  • Add to Cart