COMH4211D260 Combi-Blocks, Inc.

3-(Benzyloxy)-2-methylbenzaldehyde

  • MDL number MFCD22041841
  • Cas Number : 918524-14-8
  • Molecular Weight : 226.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9808

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9808
Availability details for
Quantity
  • Availability
  • Add to Cart