COMH4211D265 Combi-Blocks, Inc.

1-Pentanone, 5-(3r)-1,2-dithiolan-3-yl-1-[4-(2-hydroxyethyl)-1-piperazinyl]-

  • MDL number MFCD31381679
  • Cas Number : 1451180-79-2
  • Molecular Weight : 318.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9819

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9819
Availability details for
Quantity
  • Availability
  • Add to Cart