COMH4211D26B Combi-Blocks, Inc.

tert-Butyl 4-(4-amino-2-fluorophenoxy)piperidine-1-carboxylate

  • MDL number MFCD17014098
  • Cas Number : 250372-00-0
  • Molecular Weight : 310.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9841

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9841
Availability details for
Quantity
  • Availability
  • Add to Cart