COMH4211D278 Combi-Blocks, Inc.

2-[4-(4-Chloro-benzyloxy)-phenyl]-4,4,5,5-tetramethyl-[1,3,2]dioxaborolane

  • MDL number MFCD31583310
  • Molecular Weight : 344.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9862

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9862
Availability details for
Quantity
  • Availability
  • Add to Cart