COMH4211D27A Combi-Blocks, Inc.

2-Amino-3-(3-trifluoromethyl-phenyl)-propionic acid methyl ester

  • MDL number MFCD19401081
  • Cas Number : 131235-51-3
  • Molecular Weight : 247.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9871

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9871
Availability details for
Quantity
  • Availability
  • Add to Cart