COMH4211D286 Combi-Blocks, Inc.

5-Bromo-1-(cyclopropylmethyl)-1h-indole

  • MDL number MFCD27944274
  • Cas Number : 1361110-66-8
  • Molecular Weight : 250.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9888

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9888
Availability details for
Quantity
  • Availability
  • Add to Cart