COMH4211D291 Combi-Blocks, Inc.

3'-Fluoro-5'-formyl-4'-methoxybiphenyl-4-carbonitrile

  • MDL number MFCD31583171
  • Cas Number : 923281-58-7
  • Molecular Weight : 255.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9909

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9909
Availability details for
Quantity
  • Availability
  • Add to Cart