COMH4211D294 Combi-Blocks, Inc.

3-Bromo-2-(2-methoxy-ethoxy)-phenylamine

  • MDL number MFCD24118709
  • Cas Number : 1505568-38-6
  • Molecular Weight : 246.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9918

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9918
Availability details for
Quantity
  • Availability
  • Add to Cart