COMH4211D29A Combi-Blocks, Inc.

2-Amino-3-methyl-1-(4-methyl-piperazin-1-yl)-butan-1-one

  • MDL number MFCD09936496
  • Cas Number : 1097832-48-8
  • Molecular Weight : 199.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9938

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9938
Availability details for
Quantity
  • Availability
  • Add to Cart