COMH4211D29F Combi-Blocks, Inc.

[4-(4-Fluoro-benzyloxy)-pyrazol-1-yl]-acetic acid

  • MDL number MFCD31579323
  • Cas Number : 1890924-44-3
  • Molecular Weight : 250.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9944

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9944
Availability details for
Quantity
  • Availability
  • Add to Cart