COMH4211D2A1 Combi-Blocks, Inc.

[1-(Cyclopropylcarbonyl)piperidin-4-yl]acetic acid

  • MDL number MFCD16992394
  • Cas Number : 1284415-59-3
  • Molecular Weight : 211.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9946

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9946
Availability details for
Quantity
  • Availability
  • Add to Cart