COMH4211D2A5 Combi-Blocks, Inc.

Ropinirole n-oxide

  • MDL number MFCD18379620
  • Cas Number : 1076199-41-1
  • Molecular Weight : 276.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9967

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9967
Availability details for
Quantity
  • Availability
  • Add to Cart