COMH4211D2A6 Combi-Blocks, Inc.

1-[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenoxy]acetone

  • MDL number MFCD22493579
  • Cas Number : 1248556-15-1
  • Molecular Weight : 276.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9968

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9968
Availability details for
Quantity
  • Availability
  • Add to Cart