COMH4211D2A9 Combi-Blocks, Inc.

4-(6-Amino-5-benzyloxy-pyridin-3-yl)-benzoic acid methyl ester

  • MDL number MFCD31583146
  • Cas Number : 756520-51-1
  • Molecular Weight : 334.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9975

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9975
Availability details for
Quantity
  • Availability
  • Add to Cart