COMH4211D2AA Combi-Blocks, Inc.

Acetic acid, 2-[(1,3-dihydro-1,3-dioxo-2h-isoindol-2-yl)oxy]-, 2,5-dioxo-1-pyrrolidinyl ester

  • MDL number MFCD31381713
  • Cas Number : 1333377-78-8
  • Molecular Weight : 318.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9979

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9979
Availability details for
Quantity
  • Availability
  • Add to Cart