COMH4211D2AB Combi-Blocks, Inc.

2-Ethylsulfanyl-3-iodo-pyridine

  • MDL number MFCD31539534
  • Cas Number : 1643458-43-8
  • Molecular Weight : 265.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9984

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9984
Availability details for
Quantity
  • Availability
  • Add to Cart