COMH4211D2AE Combi-Blocks, Inc.

N-Ethyl-2-fluoro-4-methylaniline

  • MDL number MFCD11178357
  • Cas Number : 1040034-80-7
  • Molecular Weight : 153.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9992

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9992
Availability details for
Quantity
  • Availability
  • Add to Cart