COMH4211D2B5 Combi-Blocks, Inc.

2-Methyl-5-nitrophenyl isocyanate

  • MDL number MFCD00014718
  • Cas Number : 13471-68-6
  • Molecular Weight : 178.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QF-0008

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QF-0008
Availability details for
Quantity
  • Availability
  • Add to Cart