COMH4211D2D8 Combi-Blocks, Inc.

3,5-Bis(trifluoromethyl)-2-chloronitrobenzene

  • MDL number MFCD04972677
  • Cas Number : 654-55-7
  • Molecular Weight : 293.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QF-0832

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QF-0832
Availability details for
Quantity
  • Availability
  • Add to Cart