COMH4211D2E0 Combi-Blocks, Inc.

Sarcosine hydrochloride

  • MDL number MFCD00012609
  • Cas Number : 637-96-7
  • Molecular Weight : 125.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QF-1143

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QF-1143
Availability details for
Quantity
  • Availability
  • Add to Cart