COMH4211D31F Combi-Blocks, Inc.

2-(2-Chlorophenoxy)ethanol

  • MDL number MFCD00014034
  • Cas Number : 15480-00-9
  • Molecular Weight : 172.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QF-2563

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QF-2563
Availability details for
Quantity
  • Availability
  • Add to Cart