COMH4211D36B Combi-Blocks, Inc.

1,3-Dibromoacetone

  • MDL number MFCD00013540
  • Cas Number : 816-39-7
  • Molecular Weight : 215.9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QF-4543

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QF-4543
Availability details for
Quantity
  • Availability
  • Add to Cart