COMH4211D36C Combi-Blocks, Inc.

Trans-3-chloroacrylic acid

  • MDL number MFCD00064237
  • Cas Number : 2345-61-1
  • Molecular Weight : 106.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QF-4554

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QF-4554
Availability details for
Quantity
  • Availability
  • Add to Cart