COMH4211D378 Combi-Blocks, Inc.

2,3-Dichlorophenyl isothiocyanate

  • MDL number MFCD00041048
  • Cas Number : 6590-97-2
  • Molecular Weight : 204.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QF-4863

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QF-4863
Availability details for
Quantity
  • Availability
  • Add to Cart