COMH4211D379 Combi-Blocks, Inc.

4-(4-Chlorobenzoyl)pyridine

  • MDL number MFCD00006431
  • Cas Number : 14548-48-2
  • Molecular Weight : 217.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QF-4893

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QF-4893
Availability details for
Quantity
  • Availability
  • Add to Cart