COMH4211D392 Combi-Blocks, Inc.

4-Pyridineacetamide

  • MDL number MFCD02685124
  • Cas Number : 39640-62-5
  • Molecular Weight : 136.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QF-5491

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QF-5491
Availability details for
Quantity
  • Availability
  • Add to Cart