COMH4211D397 Combi-Blocks, Inc.

Dicyclopropylmethanol

  • MDL number MFCD00019249
  • Cas Number : 14300-33-5
  • Molecular Weight : 112.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QF-5578

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QF-5578
Availability details for
Quantity
  • Availability
  • Add to Cart