COMH4211D39F Combi-Blocks, Inc.

Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)bismuth(iii)

  • MDL number MFCD00064763
  • Cas Number : 142617-53-6
  • Molecular Weight : 758.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QF-5785

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QF-5785
Availability details for
Quantity
  • Availability
  • Add to Cart