COMH4211D3A7 Combi-Blocks, Inc.

2,4-Diamino-6-nitroquinazoline

  • MDL number MFCD00023910
  • Cas Number : 7154-34-9
  • Molecular Weight : 205.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QF-5870

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QF-5870
Availability details for
Quantity
  • Availability
  • Add to Cart