COMH4211D3BE Combi-Blocks, Inc.

Sodium heptafluorobutyrate

  • MDL number MFCD00066168
  • Cas Number : 2218-54-4
  • Molecular Weight : 236.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QF-6277

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QF-6277
Availability details for
Quantity
  • Availability
  • Add to Cart